Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006235.1 | NP_001006235.1 | 8 - 9 | 1145 | 126 | |
XM_015284619.2 | XP_015140105.1 | 10 - 11 | 1345 | 126 | ||
XM_015284625.2 | XP_015140111.1 | 9 - 10 | 1278 | 126 | ||
XM_015284631.2 | XP_015140117.1 | 9 - 10 | 1227 | 126 | ||
XM_015284637.2 | XP_015140123.1 | 8 - 9 | 1131 | 126 | ||
XM_015284646.2 | XP_015140132.1 | 7 - 8 | 1066 | 126 | ||
XM_015284657.2 | XP_015140143.1 | 6 - 7 | 970 | 126 | ||
XM_015284666.2 | XP_015140152.1 | 5 - 6 | 822 | 126 | ||
GenBank mRNA | AJ719348.1 | - | 8 - 9 | 1145 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |