Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006246.2 | NP_001006246.1 | 5 - 6 | 442 | 72 | |
XM_025148855.1 | XP_025004623.1 | 5 - 6 | 448 | 72 | ||
GenBank mRNA | AJ720085.1 | - | 5 - 6 | 442 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |