Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006262.1 | NP_001006262.1 | 5 - 6 | 544 | 117 | |
XM_015273215.2 | XP_015128701.1 | 4 - 5 | 604 | 117 | ||
XM_015273223.2 | XP_015128709.1 | 5 - 6 | 777 | 117 | ||
XM_025149757.1 | XP_025005525.1 | 4 - 5 | 600 | 117 | ||
GenBank mRNA | AJ720472.1 | - | 5 - 6 | 544 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |