Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006262.1 | NP_001006262.1 | 7 - 8 | 884 | 60 | |
XM_015273215.2 | XP_015128701.1 | 6 - 7 | 944 | 60 | ||
XM_015273223.2 | XP_015128709.1 | 7 - 8 | 1117 | 60 | ||
GenBank mRNA | AJ720472.1 | - | 7 - 8 | 884 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |