Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006264.1 | NP_001006264.1 | 3 - 4 | 378 | 110 | |
XM_015273666.2 | XP_015129152.1 | 2 - 3 | 224 | 110 | ||
GenBank mRNA | AJ720935.1 | - | 3 - 4 | 378 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |