Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARGLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006267.1 | NP_001006267.1 | 1 - 2 | 616 | 96 | |
GenBank mRNA | AJ719899.1 | - | 1 - 2 | 616 | 96 | |
CR386130.1 | - | 2 - 3 | 268 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |