Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006502.1 | NP_001006502.1 | 4 - 5 | 498 | 99 | |
GenBank mRNA | AJ720322.1 | - | 4 - 5 | 498 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |