Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006512.1 | NP_001006512.1 | 2 - 3 | 203 | 101 | |
XM_015289164.2 | XP_015144650.1 | 2 - 3 | 257 | 101 | ||
GenBank mRNA | AJ719958.1 | - | 2 - 3 | 203 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |