Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006512.1 | NP_001006512.1 | 3 - 4 | 233 | 98 | |
XM_015289164.2 | XP_015144650.1 | 3 - 4 | 287 | 98 | ||
GenBank mRNA | AJ719958.1 | - | 3 - 4 | 233 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |