Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006516.1 | NP_001006516.1 | 10 - 11 | 1219 | 98 | |
XM_025152181.1 | XP_025007949.1 | 8 - 9 | 857 | 98 | ||
GenBank mRNA | AJ719647.1 | - | 10 - 11 | 1219 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |