Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006521.2 | NP_001006521.2 | 4 - 5 | 508 | 124 | |
XM_015289800.2 | XP_015145286.1 | 4 - 5 | 522 | 124 | ||
GenBank mRNA | AJ720224.1 | - | 4 - 5 | 508 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |