Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006558.1 | NP_001006558.1 | 4 - 5 | 546 | 106 | |
XM_015291687.2 | XP_015147173.1 | 4 - 5 | 552 | 106 | ||
XM_015291688.2 | XP_015147174.1 | 5 - 6 | 638 | 106 | ||
XM_015291689.2 | XP_015147175.1 | 5 - 6 | 611 | 106 | ||
XM_015291691.2 | XP_015147177.1 | 4 - 5 | 575 | 106 | ||
XM_015291692.2 | XP_015147178.1 | 4 - 5 | 552 | 106 | ||
XM_025153395.1 | XP_025009163.1 | 4 - 5 | 552 | 106 | ||
GenBank mRNA | AJ720384.1 | - | 4 - 5 | 546 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |