Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006567.1 | NP_001006567.1 | 8 - 9 | 1085 | 143 | |
XM_015288997.2 | XP_015144483.1 | 8 - 9 | 1081 | 143 | ||
GenBank mRNA | AJ720605.1 | - | 8 - 9 | 1085 | 143 | |
BX934747.2 | - | 7 - 8 | 1047 | 143 | ||
BX935296.1 | - | 7 - 8 | 903 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |