Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006571.2 | NP_001006571.1 | 4 - 5 | 456 | 94 | |
GenBank mRNA | AJ719616.1 | - | 4 - 5 | 458 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |