Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006577.2 | NP_001006577.2 | 7 - 8 | 602 | 150 | |
XM_015277312.2 | XP_015132798.1 | 7 - 8 | 633 | 150 | ||
GenBank mRNA | AJ720892.1 | - | 7 - 8 | 603 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |