Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006577.2 | NP_001006577.2 | 9 - 10 | 940 | 72 | |
NM_001194996.1 | NP_001181925.1 | 10 - 11 | 958 | 72 | ||
XM_015277311.2 | XP_015132797.1 | 10 - 11 | 975 | 72 | ||
XM_015277312.2 | XP_015132798.1 | 9 - 10 | 971 | 72 | ||
GenBank mRNA | AJ720892.1 | - | 9 - 10 | 945 | 72 | |
BU388633.1 | - | 1 - 2 | 210 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |