Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006583.1 | NP_001006583.1 | 1 - 2 | 93 | 146 | |
GenBank mRNA | AJ719813.1 | - | 1 - 2 | 93 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |