Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007081.1 | NP_001007082.1 | 6 - 7 | 1100 | 70 | |
GenBank mRNA | AY702438.1 | - | 6 - 7 | 1100 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |