Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007084.1 | NP_001007085.1 | 3 - 4 | 319 | 153 | |
XM_015294038.1 | XP_015149524.1 | 3 - 4 | 605 | 153 | ||
GenBank mRNA | AB618613.1 | - | 3 - 4 | 289 | 153 | |
AJ621252.1 | - | 3 - 4 | 319 | 153 | ||
HM590610.1 | - | 3 - 4 | 289 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |