Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007086.1 | NP_001007087.1 | 2 - 3 | 358 | 97 | |
GenBank mRNA | BX934522.2 | - | 2 - 3 | 358 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |