Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007479.1 | NP_001007480.1 | 2 - 3 | 234 | 75 | |
GenBank mRNA | AJ720803.1 | - | 2 - 3 | 234 | 75 | |
AY389964.1 | - | 1 - 2 | 122 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |