Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007827.2 | NP_001007828.1 | 2 - 3 | 2604 | 110 | |
XM_015292000.2 | XP_015147486.1 | 3 - 4 | 3589 | 110 | ||
XM_015292001.2 | XP_015147487.1 | 4 - 5 | 3734 | 110 | ||
GenBank mRNA | AY712943.1 | - | 2 - 3 | 2608 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |