Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007828.1 | NP_001007829.1 | 10 - 11 | 1063 | 115 | |
GenBank mRNA | AJ719527.1 | - | 10 - 11 | 1063 | 115 | |
CR405973.1 | - | 5 - 6 | 554 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |