Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007847.1 | NP_001007848.1 | 3 - 4 | 401 | 60 | |
XM_015295780.1 | XP_015151266.1 | 7 - 8 | 1415 | 60 | ||
XM_015295781.2 | XP_015151267.1 | 6 - 7 | 1110 | 60 | ||
XM_015295784.2 | XP_015151270.1 | 3 - 4 | 1922 | 60 | ||
XM_015295785.2 | XP_015151271.1 | 3 - 4 | 377 | 60 | ||
XM_025141963.1 | XP_024997731.1 | 6 - 7 | 1108 | 60 | ||
XM_025141964.1 | XP_024997732.1 | 5 - 6 | 855 | 60 | ||
XM_025141965.1 | XP_024997733.1 | 4 - 5 | 662 | 60 | ||
XM_025141966.1 | XP_024997734.1 | 3 - 4 | 377 | 60 | ||
GenBank mRNA | AJ720977.1 | - | 3 - 4 | 401 | 60 | |
CR523078.1 | - | 3 - 4 | 434 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |