Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007917.2 | NP_001007918.1 | 1 - 2 | 245 | 127 | |
GenBank mRNA | AJ720634.1 | - | 1 - 2 | 210 | 127 | |
BU353194.1 | - | 1 - 2 | 249 | 127 | ||
DN929287.1 | - | 1 - 2 | 233 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |