Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007930.1 | NP_001007931.1 | 5 - 6 | 527 | 119 | |
GenBank mRNA | AJ719540.1 | - | 5 - 6 | 527 | 119 | |
CR406197.1 | - | 5 - 6 | 502 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |