Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007960.1 | NP_001007961.1 | 6 - 7 | 842 | 116 | |
GenBank mRNA | AJ720037.1 | - | 6 - 7 | 842 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |