Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008442.1 | NP_001008442.1 | 6 - 7 | 889 | 91 | |
XM_015275733.2 | XP_015131219.1 | 6 - 7 | 868 | 91 | ||
XM_015275743.2 | XP_015131229.1 | 6 - 7 | 945 | 91 | ||
XM_015275752.2 | XP_015131238.1 | 6 - 7 | 1091 | 91 | ||
XM_015275754.2 | XP_015131240.1 | 5 - 6 | 820 | 91 | ||
GenBank mRNA | AJ720668.1 | - | 6 - 7 | 889 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |