Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008453.1 | NP_001008453.1 | 525 | 86 | ||
XM_015298952.2 | XP_015154438.1 | 362 | 86 | |||
XM_015298953.2 | XP_015154439.1 | 361 | 86 | |||
GenBank mRNA | AJ720095.1 | - | 525 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus |