Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008461.1 | NP_001008461.1 | 5 - 6 | 1065 | 91 | |
XM_015284826.2 | XP_015140312.1 | 5 - 6 | 1034 | 91 | ||
XM_025148768.1 | XP_025004536.1 | 6 - 7 | 1136 | 91 | ||
GenBank mRNA | AJ720733.1 | - | 5 - 6 | 1065 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |