Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008465.1 | NP_001008465.1 | 6 - 7 | 591 | 133 | |
GenBank mRNA | AJ719495.1 | - | 6 - 7 | 591 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |