Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008469.1 | NP_001008469.1 | 5 - 6 | 565 | 163 | |
GenBank mRNA | AJ720768.1 | - | 5 - 6 | 565 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |