Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008469.1 | NP_001008469.1 | 6 - 7 | 700 | 123 | |
GenBank mRNA | AJ720768.1 | - | 6 - 7 | 700 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |