Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008475.1 | NP_001008475.1 | 4 - 5 | 320 | 85 | |
GenBank mRNA | AJ719632.1 | - | 4 - 5 | 320 | 85 | |
CR387884.1 | - | 3 - 4 | 389 | 85 | ||
CR389824.1 | - | 4 - 5 | 304 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |