Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYX1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008674.1 | NP_001008674.1 | 3 - 4 | 498 | 149 | |
GenBank mRNA | AY464141.1 | - | 3 - 4 | 498 | 149 | |
CR389904.1 | - | 3 - 4 | 476 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |