Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008683.1 | NP_001008683.1 | 8 - 9 | 2745 | 74 | |
GenBank mRNA | AY376896.1 | - | 8 - 9 | 2745 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |