Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008789.1 | NP_001008789.1 | 1 - 2 | 139 | 84 | |
GenBank mRNA | AJ812227.1 | - | 1 - 2 | 139 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |