Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204135.2 | NP_989466.1 | 3 - 4 | 247 | 83 | |
XM_015289365.2 | XP_015144851.1 | 3 - 4 | 257 | 83 | ||
GenBank mRNA | AF461015.1 | - | 3 - 4 | 255 | 83 | |
AJ719470.1 | - | 3 - 4 | 247 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |