Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204415.2 | NP_989746.2 | 1 - 2 | 123 | 100 | |
XM_025153788.1 | XP_025009556.1 | 1 - 2 | 185 | 100 | ||
GenBank mRNA | J05642.1 | - | 1 - 2 | 68 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |