Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204416.1 | NP_989747.1 | 2 - 3 | 202 | 72 | |
GenBank mRNA | M37336.1 | - | 2 - 3 | 202 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |