Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204754.1 | NP_990085.1 | 5 - 6 | 1072 | 102 | |
XM_015284009.2 | XP_015139495.1 | 5 - 6 | 1102 | 102 | ||
XM_025148524.1 | XP_025004292.1 | 5 - 6 | 1046 | 102 | ||
GenBank mRNA | D89999.1 | - | 5 - 6 | 1072 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |