Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204760.1 | NP_990091.1 | 3 - 4 | 374 | 109 | |
GenBank mRNA | AF082192.1 | - | 3 - 4 | 374 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |