Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204767.1 | NP_990098.1 | 3 - 4 | 525 | 108 | |
XM_015279317.2 | XP_015134803.1 | 3 - 4 | 822 | 108 | ||
GenBank mRNA | AF182815.1 | - | 3 - 4 | 525 | 108 | |
AF239804.1 | - | 3 - 4 | 487 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |