Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204768.1 | NP_990099.1 | 1 - 2 | 438 | 90 | |
XM_025150559.1 | XP_025006327.1 | 1 - 2 | 727 | 90 | ||
GenBank mRNA | AF178671.1 | - | 1 - 2 | 438 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |