Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204792.1 | NP_990123.1 | 3 - 4 | 848 | 121 | |
XM_015284463.2 | XP_015139949.1 | 2 - 3 | 1783 | 121 | ||
GenBank mRNA | AF152356.2 | - | 3 - 4 | 870 | 121 | |
AJ245601.1 | - | 3 - 4 | 848 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |