Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204795.1 | NP_990126.1 | 4 - 5 | 895 | 105 | |
XM_015278185.2 | XP_015133671.1 | 4 - 5 | 4081 | 105 | ||
XM_025149503.1 | XP_025005271.1 | 5 - 6 | 4168 | 105 | ||
XM_025149504.1 | XP_025005272.1 | 5 - 6 | 4153 | 105 | ||
XM_025149505.1 | XP_025005273.1 | 5 - 6 | 4168 | 105 | ||
GenBank mRNA | AF198349.1 | - | 4 - 5 | 895 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |