Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205443.2 | NP_990774.2 | 4 - 5 | 311 | 74 | |
GenBank mRNA | J05187.1 | - | 4 - 5 | 311 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |