Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205445.1 | NP_990776.1 | 1 - 2 | 130 | 75 | |
GenBank mRNA | J02828.1 | - | 1 - 2 | 130 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |