Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRFPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127170.1 | NP_001120642.1 | 5 - 6 | 898 | 129 | |
XM_015276274.2 | XP_015131760.1 | 5 - 6 | 971 | 129 | ||
GenBank mRNA | BX950747.1 | - | 2 - 3 | 895 | 129 | |
EU525171.1 | - | 5 - 6 | 898 | 129 | ||
KY856722.1 | - | 5 - 6 | 1036 | 129 | ||
KY856723.1 | - | 5 - 6 | 1036 | 129 | ||
KY856724.1 | - | 5 - 6 | 1036 | 129 | ||
KY856725.1 | - | 4 - 5 | 974 | 129 | ||
KY856726.1 | - | 3 - 4 | 815 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Gallus gallus Probe spans exons |