Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128062.1 | NP_001121534.1 | 1 - 2 | 695 | 91 | |
GenBank mRNA | EU647890.1 | - | 1 - 2 | 695 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Gallus gallus Probe spans exons |